Skip to Content
Merck
All Photos(1)

Key Documents

MAB4385

Sigma-Aldrich

Anti-TRA-1-85 Antibody, blood group Antigen Ok(a), clone TRA-1-85

clone TRA-1-85, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TRA-1-85, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

input

sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

The high-frequency blood group antigen Ok(a) is carried on a red cell membrane glycoprotein (gp) of 35-69 kDa that is widely distributed on malignant cells of different origins. Immunostaining of hemopoietic cells and a range of normal human tissues demonstrated a wide distribution of the Ok(a) gp that appears to be nonlineage-restricted, although certain tissues show differentiation-related expression. This clone can be used to monitor contamination of human Embryonic Stem cell cultures with murine feeder cells.

Specificity

This antibody exhibits pan-human reactivity, recognizing the cell surface blood group antigen Ok (a). Reactivity to murine cells has not been observed.

SPECIES REACTIVITIES:

Refer to comments regarding specificity

Immunogen

2102Ep human Embryonal Carcinoma Cells
Epitope: blood group antigen Ok(a)

Application

Anti-TRA-1-85 Antibody, blood group antigen Ok(a), clone TRA-1-85 detects level of TRA-1-85 & has been published & validated for use in FC, IP & WB.
FACS Analysis: a starting range of 5-20 μg/ml is suggested.

Immunoprecipitation

Immunoblotting

Optimal working dilutions must be determined by the end user.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Target description

35-69 kDa

Physical form

Format: Purified
Purified Immunoglobulin. Liquid in 0.02M PBS, 0.25 NaCl ph 7.6 with 0.1% Sodium Azide

Storage and Stability

Maintain refrigerated at 2-8C in undiluted aliquots for up to 12 months

Analysis Note

Control
Hemopoietic cells and a wide range of normal human tissues

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jee Myung Yang et al.
Experimental & molecular medicine, 53(4), 631-642 (2021-04-09)
Retinal degenerative disorders, including age-related macular degeneration and retinitis pigmentosa (RP), are characterized by the irreversible loss of photoreceptor cells and retinal pigment epithelial (RPE) cells; however, the long-term effect of implanting both human induced pluripotent stem cell (hiPSC)-derived RPE
Linda Harkness et al.
Methods in molecular biology (Clifton, N.J.), 873, 33-51 (2012-04-25)
In 1998, a development occurred in stem cell biology with the first report of the derivation of a human embryonic stem cell (hESC) line. Since then a number of techniques have been used to derive and characterise hESCs. Here, we
Budd A Tucker et al.
eLife, 2, e00824-e00824 (2013-08-31)
Next-generation and Sanger sequencing were combined to identify disease-causing USH2A mutations in an adult patient with autosomal recessive RP. Induced pluripotent stem cells (iPSCs), generated from the patient's keratinocytes, were differentiated into multi-layer eyecup-like structures with features of human retinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service