Skip to Content
Merck
All Photos(1)

Key Documents

MAB1954

Sigma-Aldrich

Anti-Integrin α4 Antibody, clone P4G9

ascites fluid, clone P4G9, Chemicon®

Synonym(s):

CD49d

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

P4G9, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGA4(3676)

Specificity

Human integrin alpha4. The alpha4beta1 integrin receptor recognizes an RGD-independent alternative adhesion site in the CS-1 region of fibronectin.

Application

Detect Integrin α4 using this Anti-Integrin α4 Antibody, clone P4G9 validated for use in IH.
Immunocytochemistry on Jurkat cells at 1:1,600.

Inhibits attachment of hematopoeitic cells and fibroblasts to fibronectin (note: sodium azide should be removed prior to attachment assay).

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Linkage

Replaces: 04-1131

Storage and Stability

Store at -20 °C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabienne De Toni-Costes et al.
PloS one, 5(3), e9807-e9807 (2010-03-31)
Cell survival mediated by integrin engagement has been implicated in cell adhesion-mediated drug resistance. We have recently demonstrated that the activation of glycogen synthase kinase 3 beta (GSK3beta) is a new pathway supporting the chemoresistance of leukemic cells adhered to
Barbara Garmy-Susini et al.
The Journal of clinical investigation, 115(6), 1542-1551 (2005-05-20)
Neovascularization depends on vascular cell proliferation and on the stabilization of vessels by association of vascular smooth muscle-like pericytes with ECs. Here we show that integrin alpha4beta1 (VLA-4) and VCAM-1 promote close intercellular adhesion between ECs and pericytes and that
Ming-Yi Lee et al.
PloS one, 4(10), e6913-e6913 (2009-10-23)
Human placental multipotent mesenchymal stromal cells (hPMSCs) can be isolated from term placenta, but their angiogenic ability and the regulatory pathways involved are not known. hPMSCs were shown to express integrins alpha(v), alpha(4), alpha(5), beta(1), beta(3), and beta(5) and could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service