Skip to Content
Merck

IF009

Interferon-α A Protein, Recombinant mouse

Interferon-α A Protein, Recombinant mouse.

Synonym(s):

Interferon alpha-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.75
eCl@ss:
32160405
Form:
liquid
Assay:
>95%
Biological source:
mouse
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

recombinant

expressed in E. coli

assay

>95%

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

-65 to -96°C

General description

Product Source: Gene obtained from mouse DNA expressed in E. coli.
Purified Recombinant Mouse Interferon Alpha A (IFNalphaA)

Physical form

Frozen, in PBS containing 0.1% bovine serum albumin (BSA).

Preparation Note

After receipt, this product should be kept at -70°C or below for retention of full activity. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Use polypropylene pipette tips and tubes when diluting this interferon. Dilute only in buffers containing protein such as 0.1% bovine serum albumin.

Analysis Note

Specific Activity: Determined by assaying inhibition of virus-induced cytopathic effect.In this antiviral assay for interferon, about 1 unit/mL of interferon is the quantity necessary to produce a cytopathic effect of 50%

Other Notes

Specific Activity: The units are determined with respect to the international reference standard for mouse interferon alpha (IFNalpha) provided by the National Institutes of Health.Unit of activity measured on mouse L929 cells with encephalomyocarditis virus (EMCV).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library