Skip to Content
Merck
All Photos(1)

Documents

EZFITMICAD

Millipore

EZ-Fit® adapter for using EZ-Fit® Filtration Units on Microfil® Manifolds

for using EZ-Fit® Filtration Units on Microfil® Manifolds

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

silicone

packaging

pkg of 3 units

manufacturer/tradename

EZ-Fit®

input

sample type bottled water
sample type cosmetics
sample type beer
sample type pharmaceutical(s)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

The adapter is suitable for using EZ-Fit® Filtration Units on EZ-Fit® filtration Manifolds set with Microfil® heads.

Application

EZ-Fit® adapter is a vacuum pump accessory for filtration of test samples for general microbiological analysis, bioburden testing, & water monitoring.

Preparation Note

Sterilization Method
Autoclavable

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service