Skip to Content

ECM625

IN VITRO ANGIOGENESIS ASSAY KIT

Synonym(s):

Angiogenesis assay kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161503
NACRES:
NA.41
eCl@ss:
32161000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

vertebrates

Quality Segment

packaging

pkg of 1 kit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

shipped in

dry ice

storage temp.

2-8°C

General description

The CHEMICON in vitro angiogenesis assay kit represents a simple model of angiogenesis in which the induction or inhibition of tube formation by exogenous signals can be easily monitored. Angiogenesis is the process of generating new capillary blood vessels. It is a fundamental component of several normal (reproduction and wound healing) and pathological processes (diabetic retinopathy, rheumatoid arthritis, tumor growth, and metastasis).
The CHEMICON in vitro angiogenesis assay kit provides a convenient system for evaluation of tube formation by endothelial cells in a convenient 96-well format. When cultured on ECMatrix, a solid gel of basement proteins prepared from the Engelbreth Holm-Swarm (EHS) mouse tumor, these endothelial cells rapidly align and form hollow tube-like structures. Tube formation is a multi-step process involving cell adhesion, migration, differentiation, and growth.
ECMatrix is optimized for maximal tube-formation. For assaying inhibitors or stimulators of tube formation, simply premix the endothelial cell suspension with different concentrations of the inhibitor or stimulator to be tested, before adding the cells to the top of the ECMatrix.

Application

In vitro angiogenesis assay kit has been used to monitor the extent of tube assembly/angiogenic potential in various endothelial cells such as:
  • human umbilical vein cells (HUVEC)
  • primary human retinal microvascular cells
  • platelet derived growth factor receptor-β (PDGFRβ)/PAE cells
  • bovine capillary endothelial (BCE) cells

Preparation Note

Store kit materials at -20 ºC for up to 6 months. Once ECMatrix has been thawed, store at 2-8 ºC for one week. Do not refreeze.

Other Notes

  • ECMatrixTM Gel Solution: (Part No. 90060) Five 1 mL bottles.
  • ECMatrixTM Diluent Buffer, 10X: (Part No. 90061) One 1 mL bottle.
The CHEMICON in vitro angiogenesis assay kit is intended for research use only; not for diagnostic or therapeutic applications.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library