Skip to Content
Merck

ABF192

Anti-phospho-MDA-5 (Ser104)

from rabbit, purified by affinity chromatography

Synonym(s):

Interferon-induced helicase C domain-containing protein 1, CADM-140 autoantigen, Helicase with 2 CARD domains, Helicard, Interferon-induced with helicase C domain protein 1, Melanoma differentiation-associated protein 5, MDA-5, Murabutide down-regulated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
WB, inhibition assay
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

inhibition assay: suitable (peptide), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer104)

Gene Information

human ... IFIH1(64135)

General description

Interferon-induced helicase C domain-containing protein 1 (UniProt: Q9BYX4; also known as EC: 3.6.4.13, Clinically amyopathic dermatomyositis autoantigen 140 kDa, CADM-140 autoantigen, Helicase with 2 CARD domains, Helicard, Interferon-induced with helicase C domain protein 1, Melanoma differentiation-associated protein 5, MDA-5, Murabutide down-regulated protein, RIG-I-like receptor 2, RLR-2, RNA helicase-DEAD box protein 116) is encoded by the IFIH1 (also known as MDA5, RH116) gene (Gene ID: 64135) in human. MDA-5 is an innate immune receptor that acts as a cytoplasmic sensor of viral nucleic acids and plays a major role in sensing viral infection and in the activation of a cascade of antiviral responses including the induction of type I interferons and pro-inflammatory cytokines. MDA-5 is widely expressed, but at low levels. Expression is detected at slightly higher levels in placenta, pancreas and spleen. Its ligands include mRNA lacking 2′-O-methylation at their 5′ cap and long-dsRNA. In the absence of ligands it is monomeric and homodimerizes in the presence of dsRNA ligands. Upon ligand binding it associates with mitochondria antiviral signaling protein (MAVS/IPS1), which activates the IKK-related kinases TBK1 and IKBKE that phosphorylate interferon regulatory factors IRF3 and IRF7. These factors activate transcription of antiviral immunological genes, including interferons alpha and beta. MDA-5 also plays an important role in enhancing natural killer cell function and is believed to be involved in growth inhibition and apoptosis in several tumor cell lines. MDA-5 contains 2 CARD domains; 1 helicase ATP-binding domain, 1 helicase C-terminal domain, and 1 RLR C-terminal regulatory domain.
~117 kDa observed; 116.69 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 12 amino acids surrounding Ser104 from the N-terminal region of human MDA-5.

Application

Anti-phospho-MDA-5 (Ser104), Cat. No. ABF192 is a rabbit polyclonal antibody that detects MDA-5 phosphorylated on Ser104 and has been tested for use in Western Blotting and Peptide Inhibition Assay.
Peptide Inhibition Analysis: A 1:500 dilution from a representative lot detected phospho-MDA-5 (Ser104) in Raji cell lysate.


Research Category
Inflammation & Immunology

Biochem/physiol Actions

This rabbit polyclonal antibody specifically detects human MDA-5 protein phosphorylated on Ser104.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide, pH7.4 with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in Daudi cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected phospho-MDA-5 (Ser104) in 10 µg of Daudi cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library