Skip to Content
Merck

ABD125

Anti-TFE3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

TFE3, Transcription factor E3, Class E basic helix-loop-helix protein 33, bHLHe33

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TFE3(7030)

General description

The DNA-binding factor TFE3 contains adjacent helix-loop-helix (HLH) and leucine zipper (LZ) domains flanked by an upstream basic region. These protein motifs are frequently observed in other transcription factors and are particularly common to members of the Myc family. TFE3 is ubiquitously expressed and can directly associates with DNA as either homodimers or heterodimers formed with two related proteins, TFEB or TFEC. TFE3 binds to and activates the microE3 motif of the immunoglobulin heavy-chain enhancer to induce B-cell-specific gene transcription and DNA recombination. TFEB is expressed at low levels in the embryo but at high levels in the trophoblast cells of the placenta, where it plays a critical role in regulating normal vascularization of the placenta. Recently, Smith et al. have proven that the exit from pluripotency is gated by intracellular redistribution of the bHLH transcription factor TFE3 (Cell 153, 335–347, April 11, 2013).
~ 65 kDa observed

Immunogen

linear peptide corresponding to Human TFE3.

Application

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory on TFE3 overexpressing cell lines, epiblast and primitive endodermal cells (Smith, A., et al. (2013). Cell. 153, 335–347).
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Anti-TFE3 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of TFE3.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS, 50% glycerol, and 0.05% sodium azide

Preparation Note

Store at -20°C

Analysis Note

Evaluated by Western Blot in a HEK293T cell lysate.
Western Blot Analysis: 2 µg/mL of this antibody detected TFE3 in 10 µg of HEK293T cell lysate.
Uncharacterized band(s) may appear in some lysates with longer exposure times.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library