Skip to Content
Merck
All Photos(1)

Documents

AB1510

Sigma-Aldrich

Anti-Amyloid β Antibody, aa1-14 h β-amyloid

serum, Chemicon®

Synonym(s):

Beta-APP, A-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Specificity

Beta amyloid. Sequence is contained in the Amyloid beta-A4 protein amino acids 672-685 of human APP. The sequence is identical in the following other related proteins:

Cerebral vascular amyloid peptide) (CVAP) (Protease nexin-II) (PN-II) (APPI) (PreA4) [Contains: Soluble APP-alpha (S-APP-alpha); Soluble APP-beta (S-APP-beta); C99; Beta-amyloid protein 42 (Beta-APP42); Beta-amyloid protein 40 (Beta-APP40); C83; P3(42); P3(40); Gamma-CTF(59) (Gamma-secretase C-terminal fragment 59) (Amyloid intracellular domain 59) (AID(59)); Gamma-CTF(57) (Gamma-secretase C-terminal fragment 57).

100% homology in human, pig, sheep, rabbit, bovine, 78% identities (11/14), with 12/14 positives (85%) in rat and mouse beta amyloid containing proteins.

Immunogen

Epitope: aa1-14 h beta-amyloid
Synthetic peptide that corresponds to the first 14 amino acids of the N-terminal sequence of beta-amyloid. {DAEFRHDSGYEVHH}

Application

Anti-Amyloid Antibody, β, aa1-14 h β-amyloid detects level of Amyloid & has been published & validated for use in WB, IH.
Immunohistochemistry on formic acid treated histological sections. Suggested working dilution is >1:200. AB1510 will detect both cerebrovascular and core amyloid. Formic acid pretreatment (Vinters et al., 1988) involves deparaffinizing sections, follwed by incubation in a 99% formic acid solution (10-20 min), a tap water rinse (10 min), 0.3% H202 in tris-buffered saline, then 3.0% NGS (30 min) followed by primary antibody.

Immunoblotting of formic acid extracts of brain tissue.

ELISA, direct:≥ 1:1000

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

Rabbit serum with 0.01% thimerosal.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service