Skip to Content
Merck
All Photos(2)

Documents

8.17046

SAFC

Benzalkonium chloride

(50% aqueous solution) EMPROVE® ESSENTIAL Ph Eur,NF

Pharma Manufacturing

Synonym(s):

Alkylbenzyldimethylammonium chloride (50% aqueous solution)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401

Agency

NF
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

liquid

density

0.98 g/cm3

storage temp.

15-25°C

General description

(50% aqueous solution) EMPROVE® ESSENTIAL Ph Eur,NF

Analysis Note

Assay (EP)47.5 - 52.5 %Assay (NF)95.0 - 105.0 %Identitypasses testAppearance of solutionpasses testAg (Silver)≤ 1 ppmPd (Palladium)*≤ 1 ppmSb (Antimony)≤ 9 ppmAcidic or alkaline impuritiespasses testAmines, amine saltspasses testC12 homologue (EP)≥ 40 %C14 homologue (EP)≥ 20 %Sum of C12 and C14 homologues (EP)≥ 70 %Impurity A≤ 0.5 %Impurity B≤ 0.15 %Impurity C≤ 0.05 %n-C12H25 homologne (NF)≥ 40.0 %n-C14H29 homologne (NF)≥ 20.0 %Sum of n-C12H25 and n-C14H29 homolognes (NF)≥ 70.0 %Benzyl alcohol≤ 0.5 %Benzaldehyde≤ 0.15 %(Chloromethyl)benzene≤ 0.05 %Residual solvents (ICH Q3C)excluded by the production processSulfated ash≤ 0.1 %Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).Corresponds to Ph Eur, NF

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service