Skip to Content
Merck

8.14645

Tetra-n-butylammonium chloride

greener alternative

for synthesis

Synonym(s):

Tetra-n-butylammonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H36ClN
CAS Number:
Molecular Weight:
277.92
UNSPSC Code:
12352107
EC Index Number:
214-195-7
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0%

form

solid

reaction suitability

core: ammonium

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

5-8 (20 °C, 100 g/L in H2O)

mp

41-44 °C

transition temp

flash point 110 °C

bulk density

700 kg/m3

greener alternative category

storage temp.

2-30°C

SMILES string

[Cl-].[N+](CCCC)(CCCC)(CCCC)CCCC

InChI

1S/C16H36N.ClH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

NHGXDBSUJJNIRV-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

Analysis Note

Assay (argentometric, calc. on anhydrous substance): ≥ 95.0 %
Water (K. F.): ≤ 5.0 %
Identity (IR): passes test
Might contain up to 5% bromide.
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


pictograms

Exclamation markHealth hazard

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

230.0 °F

flash_point_c

110 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library