Skip to Content
Merck
All Photos(1)

Key Documents

8.06017

Sigma-Aldrich

Formaldehyde dimethyl acetal

for synthesis

Synonym(s):

Formaldehyde dimethyl acetal, Formal, Methylal, Dimethoxymethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(OCH3)2
CAS Number:
Molecular Weight:
76.09
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-714-2
NACRES:
NA.22

vapor pressure

440 hPa ( 20 °C)

Quality Level

Assay

≥93.0% (GC)

form

liquid

expl. lim.

1.6-17.6 % (v/v)

bp

42.3 °C/1013 hPa

mp

-105 °C

transition temp

flash point -18 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(COC)C

InChI

1S/C3H8O2/c1-4-3-5-2/h3H2,1-2H3

InChI key

NKDDWNXOKDWJAK-UHFFFAOYSA-N

Application

Formaldehyde dimethyl acetal for synthesis. CAS 109-87-5, EC Number 203-714-2, chemical formula CH₂(OCH₃)₂.

Analysis Note

Assay (GC, area%): ≥ 93.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.857 - 0.859
Methanol (GC, area%): ≤ 7.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - STOT SE 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F

Flash Point(C)

-18 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service