Skip to Content
Merck
All Photos(1)

Key Documents

8.05966

Sigma-Aldrich

Isopropyl methyl ketone

for synthesis

Synonym(s):

Isopropyl methyl ketone, Methyl isopropyl ketone, 3-Methyl-2-butanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(CH3)2
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352115
EC Index Number:
209-264-3
NACRES:
NA.22
Assay:
≥98% (GC)
bp:
93-95 °C/1013 hPa
vapor pressure:
53.33 hPa ( 19.9 °C)

vapor pressure

53.33 hPa ( 19.9 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

475 °C

potency

6350 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-9 % (v/v)

refractive index

n20/D 1.388 - 1.389

bp

93-95 °C/1013 hPa

mp

-92 °C

transition temp

flash point -1 °C

solubility

8.21 g/L

density

0.80 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(C(C)C)C

InChI

1S/C5H10O/c1-4(2)5(3)6/h4H,1-3H3

InChI key

SYBYTAAJFKOIEJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.803 - 0.804
Identity (IR): passes test
Evaporation Number: 5.6

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F

Flash Point(C)

-1 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service