Skip to Content
Merck
All Photos(1)

Documents

658548

Sigma-Aldrich

AG 1478

InSolution, ≥98%, inhibitor EGF receptor tyrosine kinase

Synonym(s):

InSolution AG 1478

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14ClN3O2
CAS Number:
Molecular Weight:
315.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clearfaint tan

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C16H14ClN3O2/c1-21-14-7-12-13(8-15(14)22-2)18-9-19-16(12)20-11-5-3-4-10(17)6-11/h3-9H,1-2H3,(H,18,19,20)

InChI key

GFNNBHLJANVSQV-UHFFFAOYSA-N

General description

Highly potent and specific inhibitor of the epidermal growth factor (EGF) receptor tyrosine kinase (IC50 = 3 nM). Much higher concentrations are required for inhibition of the kinase activity of the closely related HER2 (neu/erb-B2) receptor (IC50 >100 µM), the platelet-derived growth factor (PDGF) receptor (IC50 >100 µM), and p210Bcr-Abl (IC50 >50 µM). Its mechanism of inhibition likely mimics that of the related compound, 4-(3-chloroanilino)quinazoline (CAQ), a competitive inhibitor with respect to ATP.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Epidermal growth factor receptor kinase
Product competes with ATP.
Reversible: yes
Target IC50: 3 nM against of epidermal growth factor receptor kinase

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/317 µl) solution of AG 1478 (Cat. No. 658552) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Levitski, A., and Gazit, A. 1995. Science267, 1782.
Fry, D.W., et al. 1994. Science265, 1093.
Osherov, N., and Levitski, A. 1994. Eur. J. Biochem.225, 1047.
Ward, W.H., et al. 1994. Biochem. Pharmacol. 48, 659.

Legal Information

Sold under license of U.S. Patent 5,457,105 and European Patent 0,566,266.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Giuliani et al.
Stem cell reviews and reports (2023-08-22)
In the adult mouse brain, the subventricular zone (SVZ) underlying the lateral ventricles harbours a population of quiescent neural stem cells, which can be activated (aNSCs) to initiate proliferation and generate a neurogenic lineage consisting of transit amplifying progenitors (TAPs)
Zhiguang Xiao et al.
iScience, 24(11), 103270-103270 (2021-11-12)
Neutrophil elastase (NE) promotes multiple stages of tumorigenesis. However, little is known regarding the molecular mechanisms underlying its stimulatory role. This study shows that NE triggers dose-dependent ERK signaling and cell migration in a panel of prostate cell lines representing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service