Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C28H30N4O · 2HCl
CAS Number:
Molecular Weight:
511.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
assay
≥98% (HPLC)
Quality Segment
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic), protect from light
color
white
solubility
water: 100 mM
storage temp.
2-8°C
SMILES string
Cl.Cl.N4(CCc5c(ccc(c5)C#N)C4)CC[C@@H]1CC[C@H](CC1)NC(=O)c2c3c(ncc2)cccc3
InChI
1S/C28H30N4O.2ClH/c29-18-21-5-8-23-19-32(16-13-22(23)17-21)15-12-20-6-9-24(10-7-20)31-28(33)26-11-14-30-27-4-2-1-3-25(26)27;;/h1-5,8,11,14,17,20,24H,6-7,9-10,12-13,15-16,19H2,(H,31,33);2*1H/t20-,24-;;
InChI key
HEZIOTGUXSPDAK-ONAIBGCWSA-N
General description
A brain-penetrant, high-affinity, and selective dopamine D3 receptor antagonist. High affinity for the hD3 receptor (pKi = 7.95) with 100-fold selectivity over the hD2 receptor and over 66 other receptors, enzymes, and ion channels. Rat data shows that the compound is selective for the D3 receptor over certain serotonergic and cholinergic receptors at doses up to 56.0 mg/kg and strong physiological response at 32.0 mg/kg.
Biochem/physiol Actions
Primary Target
D3
D3
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Collins, G. T. et al. 2005. J. Pharmacol. Exp. Ther.314, 310.
Reavill, C. et al. 2000. J. Pharmacol. Exp. Ther.294, 1154.
Reavill, C. et al. 2000. J. Pharmacol. Exp. Ther.294, 1154.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.