Skip to Content
Merck
All Photos(1)

Key Documents

328501

Sigma-Aldrich

ERO1 Inhibitor II, EN460

Synonym(s):

ERO1 Inhibitor II, EN460, ( Z)-2-Chloro-5-(4,5-dihydro-5-oxo-4-((5-phenyl-2-furanyl)methylene)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzoic acid, ( Z)-2-Chloro-5-(5-oxo-4-((5-phenylfuran-2-yl)methylene)-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid, (Z)-2-Chloro-5-(4,5-dihydro-5-oxo-4-((5-phenyl-2-furanyl)methylene)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzoic acid, (Z)-2-Chloro-5-(5-oxo-4-((5-phenylfuran-2-yl)methylene)-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12ClF3N2O4
Molecular Weight:
460.79
UNSPSC Code:
12352200
NACRES:
NA.28

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable thiol reactive enone (EN) compound that selectively interacts with the active-site cysteine of reduced, active form of ERO1α and inhibits its activity (IC50 = 1.9 µM). Also prevents ERO1 re-oxidation both in vitro and in mouse embryonic fibroblasts. Activates the unfolded protein response and protects ER-stressed 293T cells. Can inhibit ERO1α even in the presence of an excess amount of competing thiols. Can inhibit ERO1α even in the presence of an excess amount of competing thiols. EN460 binding to ERO1α is shown to promote the loss of flavin adenine dinucleotide (FAD) from the holoenzyme. Its inhibitory action appears to be irreversible, however addition of FAD and tris(hydroxypropyl)phosphine (Cat. No. 598250) can restore some enzyme activity.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Chu, Y., et al. 2011. Bioorg. Med. Chem. Lett.21, 1118.
Blais, J.D., et al. 2010. J. Biol. Chem.285, 20993.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brennan D Johnson et al.
ACS bio & med chem Au, 2(2), 161-170 (2022-07-28)
The flavin adenine dinucleotide containing Endoplasmic Reticulum Oxidoreductase-1 α (ERO1α) catalyzes the formation of de novo disulfide bond formation of secretory and transmembrane proteins and contributes towards proper protein folding. Recently, increased ERO1α expression has been shown to contribute to
Samira Samtleben et al.
The European journal of neuroscience, 56(8), 5177-5190 (2022-09-10)
Multiple sclerosis (MS) and its animal models are characterized by cellular inflammation within the central nervous system (CNS). The sources and consequences of this inflammation are currently not completely understood. Critical signs and mediators of CNS inflammation are reactive oxygen
Serena Germani et al.
Cell reports. Medicine, 5(3), 101439-101439 (2024-02-25)
Selenoprotein N (SEPN1) is a protein of the endoplasmic reticulum (ER) whose inherited defects originate SEPN1-related myopathy (SEPN1-RM). Here, we identify an interaction between SEPN1 and the ER-stress-induced oxidoreductase ERO1A. SEPN1 and ERO1A, both enriched in mitochondria-associated membranes (MAMs), are
Arthur Bassot et al.
Cell reports, 42(1), 111899-111899 (2023-01-01)
Endoplasmic reticulum (ER) homeostasis requires molecular regulators that tailor mitochondrial bioenergetics to the needs of protein folding. For instance, calnexin maintains mitochondria metabolism and mitochondria-ER contacts (MERCs) through reactive oxygen species (ROS) from NADPH oxidase 4 (NOX4). However, induction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service