Skip to Content
Merck

287891

5,6-Dichlorobenzimidazole 1-β-ᴅ-ribofuranoside

≥95% (HPLC), casein kinase II inhibitor, solid

Synonym(s):

5,6-Dichloro-1-β-D-ribofuranosylbenzimidazole, 5,6-Dichlorobenzimidazole Riboside, DRB, Casein Kinase II Inhibitor VII

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H12Cl2N2O4
CAS Number:
Molecular Weight:
319.14
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
eCl@ss:
32151902
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

5,6-Dichloro-1-β-D-ribofuranosylbenzimidazole, A potent, ATP-competitive, and specific inhibitor of casein kinase II (IC50 = 6 µM).

Quality Segment

description

RTECS - DD7310000

assay

≥95% (HPLC)

form

solid

potency

6 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 3 mg/mL, ethanol: 3 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

Clc1cc2[n](cnc2cc1Cl)[C@@H]3O[C@@H]([C@H]([C@H]3O)O)CO

InChI

1S/C12H12Cl2N2O4/c13-5-1-7-8(2-6(5)14)16(4-15-7)12-11(19)10(18)9(3-17)20-12/h1-2,4,9-12,17-19H,3H2/t9-,10-,11-,12-/m1/s1

InChI key

XHSQDZXAVJRBMX-DDHJBXDOSA-N

General description

A potent, ATP-competitive, and specific inhibitor of casein kinase II (IC50 = 6 µM). Reported to inhibit insulin-stimulated nuclear and cytosolic p70S6 kinase in CHO cells. Accentuates premature termination of transcription. Has been used to inhibit RNA polymerase II transcription which may be dependent on casein kinase II. Inhibits germinal vesicle breakdown in bovine oocytes. Induces G1/S cell cycle arrest.
Accentuates premature termination of transcription close to the promoter. Useful for isolation of RNA sequences near promoter sites that are responsible for mRNA formation. Useful for reversible superinduction of human fibroblast cultures. Potent, ATP-competivie and specific inhibitor of casein kinase II (IC50 = 6 µM). Has been used to inhibit RNA polymerase II transcription, which may be dependent on casein kinase II. Inhibits germinal vesicle breakdown in bovine oocytes.

Biochem/physiol Actions

Cell permeable: no
Primary Target
casein kinase II
Product competes with ATP.
Reversible: no

Preparation Note

May require warming (47°C) to achieve complete dissolution.
Room temperature (+20°C). Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Ethanol stock solutions are stable for up to 6 weeks at 4°C or for up to 3 months at -20°C.

Other Notes

Kim., S.J., and Kahn, C.R. 1997. Biochem. Biophys. Res. Commun. 234, 681.
Farin, C.E., and Yang, L. 1994. Mol. Reprod. Dev.37, 284.
Giardina, C., and Lis, J.T. 1993. J. Biol. Chem. 268, 23806.
Zandomeni, R., et al. 1986. J. Biol. Chem.261, 3414.
Mittleman, B., et al. 1983. J. Mol. Biol. 165, 461.
Zandomeni, R., et al. 1983. J. Mol. Biol. 167, 561.
Zandomeni, R., et al. 1982. Proc. Natl. Acad. Sci. USA79, 3167.
Stewart, M.W., et al. 1977. J. Gen. Virol.37, 221.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library