Skip to Content

218708

Casein Kinase II Inhibitor I

InSolution

Synonym(s):

InSolution Casein Kinase II Inhibitor I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6HBr4N3
Molecular Weight:
434.71
NACRES:
NA.77
UNSPSC Code:
12352200
Form:
liquid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable, highly selective, ATP/GTP-competitive casein kinase II (CKII) inhibitor (IC50 = 900 nM and 1.6 µM for rat liver and human recombinant CKII, respectively) and DYRK (IC50<1 µM for DYRK1a). Specificity for CKII has been reported following the screening of 33 kinases, including PKA, Akt, PKC, MKK1, ERK2, p38, JNK, Lck, Lyn, and Syk. Also reported to inhibit NS3-associated viral helicase activity in hepatitis C virus (HCV) (IC50 = 20 and 60 µM using DNA and RNA substrates, respectively) and West Nile virus (WNV) (IC50 = 1.7 µM and 900 nM using DNA and RNA substrates, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CK2 rat liver, human recombinant
Product competes with ATP.
Reversible: yes
Target IC50: 900 nM and 1.6 µM, using rat liver and human recombinant CK2, respectively

Packaging

Packaged under inert gas

Physical form

A 10 mM (5 mg/1.15 ml) solution of Casein Kinase II Inhibitor I (Cat. No. 218697 ) in DMSO.

Preparation Note

Following initial use, aliquot and refrigerate (4°C).

Analysis Note

Single spot by TLC

Other Notes

Borowski, P., et al. 2003. Eur. J. Biochem.270, 1645.
Zien, P., et al. 2003. Biochem. Biophys. Res. Commun.306, 129.
Sarno, S., et al. 2003. Biochem. J.374, 639.
Sarno, S., et al. 2001. FEBS Lett.496, 44.
Szyszka, R., et al. 1995. Biochem. Biophys. Res. Commun.208, 418."

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Carcinogenic / Teratogenic (D)


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library