Skip to Content
Merck

Skip To

1.93605

Potassium phosphate monobasic

EMPLURA®

Synonym(s):

Potassium phosphate monobasic, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
UNSPSC Code:
12352302
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

EMPLURA®

Quality Segment

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Packaging

1.93605.0521: 500G, Minimum order quantity: 20
1.93605.5021: 5 kg, Minimum order quantity: 4
1.93605.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Potassium phosphate monobasic


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters