Skip to Content
Merck

Skip To

1.93402

Acetic acid, glacial

99-100%, EMPLURA®

Synonym(s):

Acetic acid, Glacial acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
UNSPSC Code:
12352106
Beilstein/REAXYS Number:
506007
MDL number:
Assay:
99-100%
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

2.07 (vs air)

Quality Segment

product line

EMPLURA®

assay

99-100%

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F, 4 %, 59 °F

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

storage temp.

20-25°C

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Packaging

1.93402.0521: 500ML, Minimum order quantity: 20
1.93402.2521: 2.5L, Minimum order quantity: 4
1.93402.9026: 25 L, Minimum order quantity: 1
1.93402.9121: 105 L, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
137130AX0073AX0077
description

99-100%, EMPLURA®

description

EMPROVE® EXPERT, Ph. Eur., JP, USP

description

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

description

OmniTrace, ≥99%

assay

99-100%

assay

-

assay

-

assay

≥99%

Quality Level

200

Quality Level

500

Quality Level

300

Quality Level

100

storage temp.

20-25°C

storage temp.

2-25°C

storage temp.

room temp

storage temp.

room temp

refractive index

n20/D 1.371 (lit.)

refractive index

n20/D 1.371 (lit.)

refractive index

n20/D 1.371 (lit.)

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

bp

117-118 °C (lit.)

bp

117-118 °C (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

mp

16.2 °C (lit.)

mp

16.2 °C (lit.)

mp

16.2 °C (lit.)


Still not finding the right product?

Explore all of our products under Acetic acid, glacial


pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

102.2 °F - closed cup

flash_point_c

39 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library