Skip to Content
Merck

Skip To

1.37044

Ethanolamine

EMPROVE® EXPERT, Ph. Eur., BP, NF

Synonym(s):

Ethanolamine, 2-Aminoethanol, Monoethanolamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C2H7NO
CAS Number:
Molecular Weight:
61.08
UNSPSC Code:
12352104
EC Index Number:
205-483-3
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, NF, Ph. Eur.

vapor pressure

0.5 hPa ( 20 °C)

product line

EMPROVE® EXPERT

assay

98.0-100.5% (acidimetric)

form

liquid

autoignition temp.

410 °C

expl. lim.

3.4-27 % (v/v)

pH

12.1 (20 °C, 100 g/L in H2O)

kinematic viscosity

20 cSt(23 °C)

bp

171 °C/1013 hPa

mp

10.5 °C

transition temp

flash point 92.5 °C

density

1.02 g/cm3 at 20 °C

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ethanolamine, a primary amine, is typically used as a buffering agent in small molecule liquid formulations. It may also be used in large molecule downstream purification as a chromatography buffer component.
Due to its low microbial and endotoxin limits, Ethanolamine Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1371330027802400
application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

application(s)

-

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

-

agency

BP, Ph. Eur., NF

agency

NF

agency

-

agency

-

form

liquid

form

liquid

form

-

form

-

assay

98.0-100.5% (acidimetric)

assay

-

assay

≥99.5% (GC)

assay

≥99.0% (GC/NT)

product line

EMPROVE® EXPERT

product line

EMPROVE® EXPERT

product line

-

product line

-

density

1.02 g/cm3 at 20 °C

density

1.012 g/mL at 25 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Ethanolamine


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

195.8 °F - Pensky-Martens closed cup

flash_point_c

91 °C - Pensky-Martens closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters