Skip to Content
Merck

12-440

Poly (Glu4-Tyr) Peptide

lyophilized, biotin conjugate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Poly (Glu4-Tyr) Peptide, biotin conjugate,

form

lyophilized

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

shipped in

wet ice

Biochem/physiol Actions

Protein Target: Blk
Target Sub-Family: TK

Physical form

Lyophilized

Preparation Note

Lyophilized: 2 years at 4°C. Rehydrated: 6 months at -20°C

Analysis Note

Routinely evaluated as a substrate for many tyrosine kinases.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
12-44004053252475993

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service