Skip to Content
Merck
All Photos(1)

Key Documents

1.15936

Sigma-Aldrich

Erythrosine B (C.I. 45430)

for microscopy Certistain®

Synonym(s):

Erythrosine B (C.I. 45430), Tetraiodofluorescein disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86
MDL number:
UNSPSC Code:
41116130
EC Index Number:
240-474-8
Pricing and availability is not currently available.

Quality Level

form

solid

potency

1840 mg/kg LD50, oral (Rat)

composition

Dye, ≥80% gravimetric

pH

6-7 (20 °C, 10 g/L in H2O)

solubility

100 g/L

bulk density

470 kg/m3

εmax

930-1170 at 524-527 nm in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

[Na+].[Na+].Ic1c2c(cc(c1[O-])I)C(=C4C(=C(C(=O)C(=C4)I)I)O2)c3c(cccc3)C(=O)[O-]

InChI

1S/C20H8I4O5.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

IINNWAYUJNWZRM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Erythrosin B (C.I. 45430)for microscopy Certistain®, is a dry dye for microscopy that is often used as a counterstain, vizualising proteins, connective tissues, fibers and keratin in sample material of human origin. Depending on user requirements and personal preferences, it can be used for the preparation of the commonly used aqueous or alcoholic Erythrosin B staining solutions. As the dye belongs to the portfolio of Certistain® dyes, it is chemically analyzed according to strict specifications before release (CE marked and IVD product) and a high staining quality, with reliable staining results is guaranteed. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (gravimetrically): ≥ 80 %
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 524 - 527 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): 930 - 1170
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(C)

182 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service