Skip to Content
Merck

1.14402

Chromate Test Kit, colorimetric

0.005-0.10 mg/L (Cr), 0.011-0.22 mg/L (CrO₄²⁻), MQuant®

Synonym(s):

Chromate Visual Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chromate Test, colorimetric, 0.005-0.10 mg/L (Cr), 0.011-0.22 mg/L (CrO42-), for use with MQUANT®

specific analyte(s)

chromate

Quality Segment

measuring range

0.005-0.10 mg/L (Cr), 0.011-0.22 mg/L (CrO42-)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with long test tubes 0.005 - 0.01 - 0.02 - 0.03 - 0.04 - 0.05 - 0.06 - 0.08 - 0.10 mg/l Cr 0.011 - 0.022 - 0.045 - 0.07 - 0.09 - 0.11 - 0.13 - 0.18 - 0.22 mg/l CrO₄²⁻ MQuant®

Application


  • Environmental Monitoring: Chromate tests are essential for monitoring chromium(VI) levels in water and soil. These tests help in assessing environmental pollution caused by industrial discharge and are crucial for ensuring compliance with environmental regulations. Recent research has focused on the simultaneous extraction of phosphate and chromate using nano mixed (Al-Fe-Zr) oxide adsorbents, demonstrating enhanced sensitivity and accuracy in detecting chromate contamination in water (Pala et al., International Journal of Environmental Analytical Chemistry, 2021).



Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Chromate Test


pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library