Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
UNSPSC Code:
12141746
EC Index Number:
231-142-3
NACRES:
NA.22
MDL number:
Quality Segment
autoignition temp.
300-450 °C
pH
5-8 (50 g/L in H2O, suspension)
mp
1670 °C
density
4.5 g/cm3 at 20 °C
bulk density
1260 kg/m3
storage temp.
no temp limit
SMILES string
[Ti]
InChI
1S/Ti
InChI key
RTAQQCXQSZGOHL-UHFFFAOYSA-N
Analysis Note
- Identity (ICP): passes test
- Assay (cerimetric): ≥ 94.5
- Particle size (laser diffraction) (d90): ≤ 150 µm
signalword
Danger
hcodes
Hazard Classifications
Flam. Sol. 1
Storage Class
4.1B - Flammable solid hazardous materials
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
