Skip to Content

1.11132

Chloride Test Kit, titrimetric

titrimetric, for use with MCOLORTEST®

Synonym(s):

Chloride Visual Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chloride Test, titrimetric, for use with MCOLORTEST®

specific analyte(s)

chloride

Quality Segment

compatibility

for use with MCOLORTEST®

detection method

titrimetric

storage temp.

15-25°C

General description

Chloride is found in the earth´s crust and can therefore also be found in all natural waters at some concentration. This MQuant® Chloride test (formally MColortest®) is suitable for the titrimetric determination using a convenient dropping bottle.

Method
In nitric solution chloride ions are titrated with mercury(II) nitrate solution against 1,5-diphenylcarbazone as the indicator, forming slightly dissociated mercury(II) chloride in the process. At the titration end-point, excess mercury(II) ions react with the indicator to form a blue-violet complex. The chloride concentration is determined from the consumption of titration solution (mercurimetric determination).

The test can be used in
  • Groundwater, surface water, and seawater (after dilution)
  • Waters from aquaculture
  • Drinking water and mineral water
  • Water for concrete in the construction industry
  • Industrial water and wastewater
  • Boiler and boiler feed water, cooling water
  • Swimming-pool water
  • Food after appropriate sample pretreatment
  • Soils and fertilizers after appropriate sample pretreatment

This test is suitable to detect chloride interferences which are a mayjor reason for wron COD readings. Therefore it can serve as pretest for COD measurements with e.g. our Spectroquant® COD cell tests for seawater/samples with high chloride content 1.17058 and 1.17059.
Method: titrimetric with dropping bottle MQuant®

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Still not finding the right product?

Explore all of our products under Chloride Test


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B - STOT RE 2

target_organs

Kidney

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library