870333P
Avanti
C2:0 L-carnitine-d9
Avanti Research™ - A Croda Brand
Synonym(s):
Acetyl L-carnitine-d9
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C9H8D9NO4
CAS Number:
Molecular Weight:
212.29
UNSPSC Code:
12352211
NACRES:
NA.21
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 x 1 mg
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−20°C
Application
Lipidomics
Packaging
5 mL Amber Glass Screw Cap Vial (870333P-1MG)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service