Z531847
Tubing adapter with Torion™ connection and stopcock
PTFE stopcock, O.D. 3/16 in., male joint: ST/NS 19/22
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Recommended Products
material
PTFE stopcock
Joint
male joint: ST/NS19/22
O.D.
3/16 in.
4.8 mm
General description
PTFE stopcocks are threaded for additional safety and have a 2 mm bore for 3/16 in. tubing connection and 4 mm bore for 5/16 in. and 1/2 in. tubing connections. 5/16in. O.D fits a #8 Torion, and 1/2in. O.D. fits a #14 Torion.
Legal Information
Torion is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)