Key Documents
Z529028
Sigma-Aldrich
VerSA-Flow can and drum dispenser
Economy pressure-delivery cap
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Related Categories
General description
For low-flow pressure-delivery of solvents and for waste solvent collection using Aldrich cans and drums with tapered 2 inch NPT thread. Use at <5 psi max.
  • Solid machined PBT body
  • Includes 10 ft × 1/4 in. O.D. FEP tubing, adjustable depth through compression fittings on cap
  • Can be adapted to 2 in. buttress threads with optional thread adapter Z529036
Please refer to Aldrich Technical Information Bulletin AL-210 for additional VerSA-Flow can and drum dispensers and details for using this equipment. This bulletin is available from the Technical Reference section on our website at www.sigma-aldrich.com
Legal Information
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)