Z267600
Round crystal window
CsI, diam. × thickness 25 mm × 4 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
CsI
technique(s)
IR spectroscopy: suitable
diam. × thickness
25 mm × 4 mm
transmittance
40,000-20,000 (wavenumbers (cm-1))
refractive index
n/D 1.74 (at 2 μm)
Looking for similar products? Visit Product Comparison Guide
General description
A soft crystal; soluble in water; hygroscopic; offers an extended transmission range.
Cut and polished quality crystal windows.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service