Key Documents
SYNP150100M
Synthware synthesis vessel with lower joint and PTFE T-bore stopcock
100 mL, joint: ST/NS 24/40, porosity: medium, with drip tip
Synonym(s):
Synthware P150100M, Solid phase peptide synthesis vessel
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P150100M
capacity
100 mL
100 mL
Related Categories
General description
Solid phase peptide synthesis vessels feature 2 mm PTFE stopcocks and fritted glass supports for resin beads during operation. The 24/40 inner drip joint can be assembled with other standard glassware. The vessel has a 8 mm hose connection for vacuum and the stopcock has an 8 mm O.D. sidearm for inert gas introduction. Vessels are supplied with PTFE-faced silicone lined Nylon cap.
Legal Information
Synthware is a trademark of Kemtech America Inc.