Key Documents
SYNF609600C
Synthware Buchner funnel
600 mL, porosity: coarse
Synonym(s):
Synthware F609600C, filtering funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F609600C
capacity
600 mL
600 mL
General description
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)