Merck
SYNF301950
Synthware single-neck round-bottom flask
50 mL, joint: ST/NS 19/22
Synonym(s):
Synthware F301950, glass round flask
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121804
Capacity:
50 mL, 50 mL
Material:
borosilicate glass flask, round bottom , single-neck
Joint:
joint: ST/NS19/22
Packaging:
pkg of 1 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
borosilicate glass flask, round bottom , single-neck
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F301950
capacity
50 mL, 50 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)