Skip to Content
Merck

GF08954792

Inconel® alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi

rod, 200mm x 20mm diameter, annealed

Synonym(s):

NI137936

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
11101712
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

rod

reaction suitability

core: chromium, core: iron, core: molybdenum, core: nickel, core: niobium, core: titanium

manufacturer/tradename

Goodfellow 089-547-92

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library