Skip to Content
Merck
All Photos(2)

Key Documents

901762

Sigma-Aldrich

Tris(dimethylamino)sulfonium bifluoride

greener alternative

Synonym(s):

(Me2N)3S+[FHF], Bifluoride salt SuFEx catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H19F2N3S
CAS Number:
Molecular Weight:
203.30
UNSPSC Code:
12352101
form:
powder or solid

form

powder or solid

reaction suitability

reagent type: catalyst
reaction type: Fluorinations

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Bifluoride salt reported by K. Barry Sharpless and colleagues as a powerful catalyst for the sulfur(VI) fluoride exchange (SuFEx) reaction between aryl silyl ethers and aryl fluorosulfates or alkyl sulfonyl fluorides. Superior to organosuperbases DBU and BEMP, tris(dimethylamino)sulfonium bifluoride enables low catalyst loading (0.05 mol%) for preparation of polysulfates and polysulfonates with high molecular weight and narrow polydispersity. Reaction is amenable to scale up.

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Wang et al.
Angewandte Chemie (International ed. in English), 56(37), 11203-11208 (2017-08-10)
The SuFEx-based polycondensation between bisalkylsulfonyl fluorides (AA monomers) and bisphenol bis(t-butyldimethylsilyl) ethers (BB monomers) using [Ph

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service