714771
Glyceryl tri(oleate-1-13C)
endotoxin tested, 99 atom % 13C, 98% (CP)
Synonym(s):
Triolein-carboxyls-13C3
About This Item
Recommended Products
isotopic purity
99 atom % 13C
Assay
98% (CP)
form
liquid
bp
235-240 °C/18 mmHg
density
0.913 g/mL at 25 °C
suitability
endotoxin tested
mass shift
M+3
storage temp.
−20°C
SMILES string
CCCCCCCC\C=C/CCCCCCC[13C](=O)OCC(CO[13C](=O)CCCCCCC\C=C/CCCCCCCC)O[13C](=O)CCCCCCC\C=C/CCCCCCCC
InChI
1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-/i55+1,56+1,57+1
InChI key
PHYFQTYBJUILEZ-UBEHWWNBSA-N
Application
Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.
Packaging
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service