68070
2-Methylnonane
≥98.5% (GC)
Synonym(s):
2-Methylnonane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)6CH(CH3)2
CAS Number:
Molecular Weight:
142.28
Beilstein:
1696990
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98.5% (GC)
refractive index
n20/D 1.410
bp
166-169 °C (lit.)
density
0.726 g/mL at 20 °C (lit.)
SMILES string
CCCCCCCC(C)C
InChI
1S/C10H22/c1-4-5-6-7-8-9-10(2)3/h10H,4-9H2,1-3H3
InChI key
SGVYKUFIHHTIFL-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
114.8 °F - closed cup
Flash Point(C)
46 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service