Skip to Content
Merck
All Photos(1)

Documents

678333

Sigma-Aldrich

2-(Tributylstannyl)pyridine

85%

Synonym(s):

(2-Pyridinyl)tributylstannane, 2-(Tributyltin)pyridine, 2-Pyridyltri-n-butyltin, Bu3SnPy, Tributyl-2-pyridyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H4NSn((CH2)3CH3)3
CAS Number:
Molecular Weight:
368.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

85%

density

1.137 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccccn1

InChI

1S/C5H4N.3C4H9.Sn/c1-2-4-6-5-3-1;3*1-3-4-2;/h1-4H;3*1,3-4H2,2H3;

InChI key

GYUURHMITDQTRU-UHFFFAOYSA-N

Application

Building block in an efficient, palladium-catalyzed synthesis of 2-pyridylazaazulene, a bidentate ligand showing pH and cationic-metal dependent emission spectra.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.0 °F - closed cup

Flash Point(C)

23.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 48, 4471-4471 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service