654914
Nitrilotriacetic acid-d9
98 atom % D, 98% (CP)
Synonym(s):
N,N-bis[(carboxy-d)methyl-d2]-glycine-1,2,2-d3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
N(CD2COOD)3
CAS Number:
Molecular Weight:
200.19
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Recommended Products
isotopic purity
98 atom % D
Assay
98% (CP)
form
solid
mp
245 °C (lit.)
mass shift
M+9
SMILES string
[2H]OC(=O)C([2H])([2H])N(C([2H])([2H])C(=O)O[2H])C([2H])([2H])C(=O)O[2H]
InChI
1S/C6H9NO6/c8-4(9)1-7(2-5(10)11)3-6(12)13/h1-3H2,(H,8,9)(H,10,11)(H,12,13)/i1D2,2D2,3D2/hD3
InChI key
MGFYIUFZLHCRTH-HCHPEDFLSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service