445916
Ethyl vinyl sulfide
96%
Synonym(s):
(Ethylthio)ethene, Ethyl vinyl sulfide, Ethylsulfanylethene, Vinyl ethyl sulfide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C2H5SCH=CH2
CAS Number:
Molecular Weight:
88.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
96%
form
liquid
refractive index
n20/D 1.478 (lit.)
bp
91-92 °C (lit.)
density
0.869 g/mL at 25 °C (lit.)
SMILES string
CCSC=C
InChI
1S/C4H8S/c1-3-5-4-2/h3H,1,4H2,2H3
InChI key
AFGACPRTZOCNIW-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
21.2 °F - closed cup
Flash Point(C)
-6 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
D R Warner et al.
Biochemistry, 35(14), 4480-4484 (1996-04-09)
Thioether S-methyltransferase is an important enzyme in the metabolism of sulfur and selenium-containing compounds in animals. Ethyl vinyl sulfide was previously shown to be a substrate for this enzyme yielding methyl ethyl vinyl sulfonium ion (MEVS+) upon reaction with S-adenosylmethionine.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service