Skip to Content
Merck
All Photos(1)

Key Documents

309982

Sigma-Aldrich

Ethyl (R)-(−)-mandelate

99%, optical purity ee: 99% (GLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CO2C2H5
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

optical activity

[α]21/D −134°, c = 3 in chloroform

optical purity

ee: 99% (GLC)

bp

103-105 °C/2 mmHg (lit.)

mp

33-34 °C (lit.)

SMILES string

CCOC(=O)[C@H](O)c1ccccc1

InChI

1S/C10H12O3/c1-2-13-10(12)9(11)8-6-4-3-5-7-8/h3-7,9,11H,2H2,1H3/t9-/m1/s1

InChI key

SAXHIDRUJXPDOD-SECBINFHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asteriani R Dewanti et al.
Biochemistry, 43(7), 1883-1890 (2004-02-18)
(S)-Mandelate dehydrogenase (MDH) from Pseudomonas putida is a flavin mononucleotide (FMN)-dependent enzyme that oxidizes (S)-mandelate to benzoylformate. In this work, we show that the ethyl and methyl esters of (S)-mandelic acid are substrates for MDH. Although the binding affinity of
H Xiao et al.
Chinese journal of biotechnology, 9(1), 33-39 (1993-01-01)
Candida cyclindracea lipase (CCL) was added to "sodium dodecyl sulfonate (AS)/n-butanol/n-octane/n-octane" water-in-oil microemulsion to catalyze the hydrolysis of ethyl mandelate and the esterification of alpha-bromopropionic acid with n-butanol, respectively. The catalytic activity of CCL in the above microemulsions was higher

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service