Merck
  • Capillary electrophoresis with field-amplified sample stacking for rapid and sensitive determination of sulfadiazine and sulfamethoxazole.
Capillary electrophoresis with field-amplified sample stacking for rapid and sensitive determination of sulfadiazine and sulfamethoxazole.
Die Pharmazie (2012-09-29)
Yuqin Li, Yingjie Cui, Baoxiu Jia, Hao Wang, Caihong Liu, Yongxiu Qi
ABSTRACT
A new capillary electrophoresis method with field-amplified sample stacking (FASS) was developed for the analysis of sulfadiazine and sulfamethoxazole. After optimization of the separation and concentration conditions, the two compounds can be separated within 7 min and quantified with high sensitivity, with detection limits of 0.48 ng/mL for sulfadiazine and 0.76 ng/mL for sulfamethoxazole. This resulted in a 300-1500-fold improvement in concentration sensitivity relative to conventional capillary electrophoresis methods. The method was useful for qualitative and quantitative analysis of sulfadiazine and sulfamethoxazole in their preparations with recovery of 99.0%-102% for sulfadiazine and 99.5% - 99.7% for sulfamethoxazole.
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Supelco
Sulfamethoxazole
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material