Merck
  • MicroRNA-148b-3p is involved in regulating hypoxia/reoxygenation-induced injury of cardiomyocytes in vitro through modulating SIRT7/p53 signaling.
MicroRNA-148b-3p is involved in regulating hypoxia/reoxygenation-induced injury of cardiomyocytes in vitro through modulating SIRT7/p53 signaling.
Chemico-biological interactions (2018-10-12)
Meng Sun, Mengen Zhai, Nan Zhang, Rui Wang, Hongliang Liang, Qinghua Han, Yongping Jia, Liqin Jiao
ABSTRACT
Accumulating evidence has suggested that microRNAs (miRNAs) are emerging as critical regulators in myocardial ischemia/reperfusion injury. miR-148b-3p has been reported to regulate cell apoptosis of various cell types. However, whether miR-148b-3p is involved in regulating cardiomyocyte apoptosis in myocardial ischemia/reperfusion injury remains unknown. In this study, we aimed to investigate the potential role and molecular mechanism of miR-148b-3p in regulating cardiomyocyte apoptosis induced by hypoxia/reoxygenation (H/R) injury in vitro, a cellular model of myocardial ischemia/reperfusion injury. We found that miR-148b-3p expression was significantly up-regulated in response to H/R treatment in cardiomyocytes. Functional experiments showed that miR-148b-3p overexpression significantly decreased the viability, increased LDH release and promoted the apoptosis of H/R-treated cardiomyocytes. In contrast, miR-148b-3p inhibition improved the viability, decreased LDH release and reduced the apoptosis of H/R-treated cardiomyocytes, showing a protective effect against H/R-induced injury. Bioinformatics analysis predicted that Sirtuin7 (SIRT7), a critical stress survival gene of cardiomyocytes, was a potential target gene of miR-148b-3p, which was then validated by dual-luciferase reporter assay, real-time quantitative polymerase chain reaction and Western blot analysis. Moreover, our results showed that miR-148b-3p regulated the acetylation of the p53 protein and modulated p53-mediated pro-apoptotic signaling through targeting SIRT7. Notably, the silencing of SIRT7 significantly abrogated miR-148b-3p inhibition-mediated cardio-protective effects, while SIRT7 overexpression rescued miR-148b-3p-induced cell apoptosis in cardiomyocytes with H/R treatment. Overall, our results indicate that miR-148b-3p contributes to the regulation of H/R-induced cardiomyocyte apoptosis in vitro through targeting SIRT7 and modulating p53-mediated pro-apoptotic signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human SIRT7
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder