Skip to Content
Merck
All Photos(1)

Key Documents

EHU108041

Sigma-Aldrich

MISSION® esiRNA

targeting human SIRT7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAGTGTGGACACTGCTTCAGAAAGGGAGAAGCGTTAGTGCTGCCGACCTGAGCGAGGCCGAGCCACTCACCCACATGAGCATCACCCGTCTGCATGAGCAGAAGCTGGTGCAGCATGTGGTGTCTCAGAACTGTGACGGGCTCCACCTGAGGAGTGGGCTGCCGCGCACGGCCATCTCCGAGCTCCACGGGAACATGTACATTGAAGTCTGTACCTCCTGCGTTCCCAACAGGGAGTACGTGCGGGTGTTCGATGTGACGGAGCGCACTGCCCTCCACAGACACCAGACAGGCCGGACCTGCCACAAGTGTGGGACCCAGCTGCGGGACACCATTGTGCACTTTGGGGAGAGGGGGACGTTGGGGCAGCCTTTGAACTGGGAAGCGACCGAGGCTGCCAGCAGAGCAGACACCATCCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Jia et al.
IUBMB life, 73(1), 264-272 (2020-12-17)
Oral squamous cell carcinoma (OSCC) is a common malignant cancer with unfavorable prognosis, and the epithelial-to-mesenchymal transition (EMT) is a critical contributor to OSCC metastasis. Recently, we have shown that sirtuin 7 (Sirt7) is associated with EMT and OSCC metastasis
Romain Haider et al.
Oncotarget, 8(44), 77309-77316 (2017-11-05)
Predictive biomarkers for advanced prostate cancer (PCa) are still missing. The sirtuin 7 (SIRT7) has been linked to tumorogenesis but its role in prostate cancer is poorly documented. To determine if SIRT7 can be a biomarker for aggressive prostate cancer
Anne E Wyman et al.
American journal of physiology. Lung cellular and molecular physiology, 312(6), L945-L958 (2017-04-08)
Pulmonary fibrosis is a severe condition with no cure and limited therapeutic options. A better understanding of its pathophysiology is needed. Recent studies have suggested that pulmonary fibrosis may be driven by accelerated aging-related mechanisms. Sirtuins (SIRTs), particularly SIRT1, SIRT3
Wang Wei et al.
American journal of cancer research, 7(9), 1788-1803 (2017-10-06)
It is still a controversy whether the role of Sirtuin 7 (SIRT7) is an oncogene or a tumor suppressor gene in cancer as SIRT7 may have different functions in different types of cancer. Particularly, the specific roles of SIRT7 in
Yingying Zhao et al.
Oncology reports, 44(3), 959-972 (2020-07-25)
Increasing evidence has indicated the roles of sirtuin 7 (SIRT7) in numerous human cancers. However, the effects and the clinical significance of SIRT7 in human lung cancer is largely unknown. The present research demonstrated that SIRT7 was increased in human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service