Skip to Content
Merck
All Photos(1)

Key Documents

V800395

Sigma-Aldrich

Sodium oxalate

LR, ≥99.55%

Synonym(s):

Ethanedioic acid sodium salt, Oxalic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein:
3631622
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥99.55%

form

crystalline powder

SMILES string

[Na+].[Na+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

ZNCPFRVNHGOPAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiří Jan et al.
Water research, 47(2), 547-557 (2012-12-12)
Correct identification of P forms together with their main Fe and Al binding partners in non-calcareous sediments is of crucial importance for evaluation of P cycling in water bodies. In this paper, we assess extraction methods frequently used for this
Juntaek Oh et al.
The Journal of biological chemistry, 289(16), 11465-11475 (2014-03-13)
The Burkholderia species utilize acetyl-CoA and oxaloacetate, substrates for citrate synthase in the TCA cycle, to produce oxalic acid in response to bacterial cell to cell communication, called quorum sensing. Quorum sensing-mediated oxalogenesis via a sequential reaction by ObcA and
Lu Gao et al.
International journal of phytoremediation, 15(4), 307-319 (2013-03-16)
Phytolacca americana L. can accumulate large amounts of heavy metals in its aerial tissues, especially cadmium (Cd) and manganese (Mn). It has great potential for use in phytoextraction of metals from multi-metal-contaminated soils. This study was conducted to further investigate
Taiying Zhang et al.
Carbohydrate polymers, 92(1), 334-344 (2012-12-12)
Dilute oxalic acid pretreatment was applied to maple wood to improve compatibility with downstream operations, and its performance in pretreatment and subsequent enzymatic hydrolysis was compared to results for hydrothermal and dilute hydrochloric and sulfuric acid pretreatments. The highest total
Sindhu Jagadamma et al.
PloS one, 7(11), e50434-e50434 (2012-12-05)
Physico-chemical sorption onto soil minerals is one of the major processes of dissolved organic carbon (OC) stabilization in deeper soils. The interaction of DOC on soil solids is related to the reactivity of soil minerals, the chemistry of sorbate functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service