Skip to Content
Merck
All Photos(1)

Key Documents

V001453

Sigma-Aldrich

1,2-Propanediol

USP, 99.5%

Synonym(s):

Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
PubChem Substance ID:

Agency

USP

Quality Level

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

99.5%

autoignition temp.

779 °F

expl. lim.

12.5 %

refractive index

n20/D 1.432 (lit.)

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service