Skip to Content
Merck
All Photos(1)

Key Documents

1295662

USP

beta-Glycerylphosphorylcholine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1,3-Dihydroxypropan-2-yl 2-(trimethylammonio)ethyl phosphate)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
3.530,00 kr.

3.530,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
3.530,00 kr.

About This Item

Empirical Formula (Hill Notation):
C8H20NO6P
CAS Number:
Molecular Weight:
257.22
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

3.530,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

Agency

USP

manufacturer/tradename

USP 1295662

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service