1112423
USP
Indomethacin Related Compound B
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
4-Chlorobenzoic acid
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
indomethacin
manufacturer/tradename
USP
mp
238-241 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
OC(=O)c1ccc(Cl)cc1
InChI
1S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)
InChI key
XRHGYUZYPHTUJZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also, for use with USP monographs such as:
- Indomethacin
- Indomethacin Capsules
- Indomethacin Sodium
- Indomethacin for Injection
- Indomethacin Oral Suspension
- Indomethacin Suppositories
- Indomethacin Extended-Release Capsules
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
460.4 °F - closed cup
Flash Point(C)
238 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service