Skip to Content
Merck
All Photos(1)

Key Documents

TA9H93ED7668

Targetmol Chemicals Inc.

KR-33493

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
460.35

Iupac Name

4-(2-((4-bromophenyl)thio)acetamido)-1-phenethyl-1H-pyrazole-3-carboxylic acid

Inchi Code

1S/C20H18BrN3O3S/c21-15-6-8-16(9-7-15)28-13-18(25)22-17-12-24(23-19(17)20(26)27)11-10-14-4-2-1-3-5-14/h1-9,12H,10-11,13H2,(H,22,25)(H,26,27)

InChI key

ZRLJEHIUGYTTSZ-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

99.71%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/KR-33493

MSDS

https://www.targetmol.com/compound/KR-33493


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library