Skip to Content
Merck
All Photos(1)

Documents

SY3H3D67587F

Synthonix Corporation

2-Amino-6-bromo-3-chloro-5-fluorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
268.47
MDL number:

Iupac Name

2-amino-6-bromo-3-chloro-5-fluorobenzoic acid

Inchi Code

1S/C7H4BrClFNO2/c8-5-3(10)1-2(9)6(11)4(5)7(12)13/h1H,11H2,(H,12,13)

InChI key

WRCZGYIVEVFSBI-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

White to light-yellow powder or crystals

Product Link

https://synthonix.com/44450.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://synthonix.com/44450.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service