Key Documents
SU861027
Supelco
Super Clean (Base-Plate Design) Gas Purifier
triple trap (hydrocarbon, moisture, oxygen), for helium carrier gas
Select a Size
1 EA
2.880,00 kr.
2.880,00 kr.
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
2.880,00 kr.
About This Item
UNSPSC Code:
41115717
NACRES:
SB.54
2.880,00 kr.
Please contact Customer Service for Availability
Request a Bulk Order
Quality Level
General description
Gas purifier helps in enhancing the performance of GC detectors. It can reduce the background signal in flame ionization and nitrogen phosphorus detectors.[1]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Gas Chromatographic Techniques and Applications
Alan John Handley, E. R. Adlard
Science, 135-135 (2001)